Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Hydroxyhippuric acid, 98%, Thermo Scientific™

Catalog No. AC208050250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC208050250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC208050250 Supplier Thermo Scientific Chemicals Supplier No. 208050250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16555-77-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00004228
InChI Key GCWCVCCEIQXUQU-UHFFFAOYSA-N
Synonym alpha-hydroxyhippuric acid, 2-benzoylamino-2-hydroxyacetic acid, alpha-hydroxybenzoylglycine, benzamidohydroxyacetic acid, 2-hydroxy-2-phenylcarbonylamino acetic acid, .alpha.-hydroxyhippuric acid, hydroxyhippuric acid, acetic acid, benzoylamino hydroxy, a-hydroxyhippuric acid, pubchem13888
PubChem CID 450272
ChEBI CHEBI:68451
IUPAC Name 2-benzamido-2-hydroxyacetic acid
SMILES C1=CC=C(C=C1)C(=O)NC(C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H5CONHCH(OH)CO2H
Quantity 25 g
Beilstein 09, I, 116
Formula Weight 195.17
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material α-Hydroxyhippuric acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.