accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16563-14-7

IUPAC Name: 4-oxothiolane-3-carbonitrile
Synonyms: 4-oxothiolane-3-carbonitrile
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: OHPSBDAUCJNDHP-UHFFFAOYNA-N
SMILES: O=C1CSCC1C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results