accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16570-28-8

IUPAC Name: dicopper(2+) (phosphonatooxy)phosphonate
Synonyms: cupric pyrophosphate
Molecular Weight (g/mol): 301.03
Molecular Formula: Cu2O7P2
InChi Key: PEVJCYPAFCUXEZ-UHFFFAOYSA-J
SMILES: [Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)