Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) pyrophosphate hydrate

Catalog No. 344699500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-469-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-469-9500G Supplier Sigma Aldrich Supplier No. 344699500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(II) pyrophosphate hydrate was one of the constituents in the preparation of an electrolyte which was used to study the application of nickel-tin (Ni-Sn) nanowires as electrodes for Li-ion batteries.

Specifications

CAS 16570-28-8
Molecular Formula Cu2P2O7 -+ xH2O
Linear Formula Cu2P2O7 -+ xH2O
Quantity 500 g
MDL Number MFCD00150225
Synonym Cupric pyrophosphate hydrate
Molecular Weight (g/mol) 301.04
Percent Purity 33-35%Cu basis (EDTA titration)
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.