accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 1658-08-8

IUPAC Name: 1,2,3,4,5,6,7,8-octahydroacridine
Synonyms: 1,2,3,4,5,6,7,8-octahydroacridine
Molecular Weight (g/mol): 187.29
Molecular Formula: C13H17N
InChi Key: LLCXJIQXTXEQID-UHFFFAOYSA-N
SMILES: C1CCC2=NC3=C(CCCC3)C=C2C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results