Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4,5,6,7,8-Octahydroacridine 97.0+%, TCI America™
SDP

Catalog No. O01975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01975G Supplier TCI America Supplier No. O01975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1658-08-8
Molecular Formula C13H17N
Molecular Weight (g/mol) 187.286
MDL Number MFCD00005032
InChI Key LLCXJIQXTXEQID-UHFFFAOYSA-N
PubChem CID 74263
IUPAC Name 1,2,3,4,5,6,7,8-octahydroacridine
SMILES C1CCC2=NC3=C(CCCC3)C=C2C1

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 187.29
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,5,6,7,8-Octahydroacridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.