accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFF

CAS RN 16583-06-5

IUPAC Name: 2,3,4,5-tetrafluorobenzaldehyde
Synonyms: 2,3,4,5-tetrafluorobenzaldehyde
Molecular Weight (g/mol): 178.09
Molecular Formula: C7H2F4O
InChi Key: UPJHEKIKCNDMEX-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results