Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. T18975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18975G 5 g
T18971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18975G Supplier TCI America Supplier No. T18975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16583-06-5
Molecular Formula C7H2F4O
Molecular Weight (g/mol) 178.086
MDL Number MFCD02683041
InChI Key UPJHEKIKCNDMEX-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3,4,5-tetrafluoro, tetrafluorobenzaldehyde, pubchem1465, acmc-209dtb, intermediates-zcf02244, 2,3,4,5-tetafluorobenzaldehyde, 2,3,4,5-tetrafluoro-benzaldehyde
PubChem CID 2734023
IUPAC Name 2,3,4,5-tetrafluorobenzaldehyde
SMILES C1=C(C(=C(C(=C1F)F)F)F)C=O

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 1989
Formula Weight 178.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5-Tetrafluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.