accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(S)OO(R)H3COHOO

CAS RN 16597-50-5

IUPAC Name: benzyl (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms: benzyl (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate
Molecular Weight (g/mol): 343.38
Molecular Formula: C19H21NO5
InChi Key: VBKUVUJWFDXTMS-PBHICJAKSA-N
SMILES: C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results