Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-threonine Benzyl Ester 98.0+%, TCI America™
SDP

Catalog No. C22855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22855G Supplier TCI America Supplier No. C22855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16597-50-5
Molecular Formula C19H21NO5
Molecular Weight (g/mol) 343.38
MDL Number MFCD00077041
InChI Key VBKUVUJWFDXTMS-PBHICJAKSA-N
Synonym z-thr-obzl, n-cbz-l-threonine benzyl ester, cbz-l-threonine benzyl ester, 2s,3r-benzyl 2-benzyloxy carbonyl amino-3-hydroxybutanoate, n-benzyloxycarbonyl-l-threonine benzyl ester, l-threonine, n-phenylmethoxy carbonyl-, phenylmethyl ester, z-l-threonine alpha-benzyl ester, benzyl 2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanoate, phenylmethyl 2s,3r-3-hydroxy-2-phenylmethoxy carbonylamino butanoate, z-l-threonine benzyl ester
PubChem CID 7018808
IUPAC Name benzyl (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate
SMILES C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 5 g
Formula Weight 343.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-threonine Benzyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.