accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OO(S)H3N

CAS RN 166023-31-0

IUPAC Name: (2S)-4-(tert-butoxy)-4-oxo-1-phenylbutan-2-aminium
Synonyms: (2S)-4-(tert-butoxy)-4-oxo-1-phenylbutan-2-aminium
Molecular Weight (g/mol): 236.33
Molecular Formula: C14H22NO2
InChi Key: ZIJHIHDFXCNFAA-LBPRGKRZSA-O
SMILES: CC(C)(C)OC(=O)C[C@@H]([NH3+])CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)