Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3R)-3-amino-4-phenylbutanoate 97%, Thermo Scientific™

Catalog No. AC344451000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC344451000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC344451000 Supplier Thermo Scientific Chemicals Supplier No. 344451000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166023-31-0
Molecular Formula C14H22NO2
Molecular Weight (g/mol) 236.33
MDL Number MFCD00798307
InChI Key ZIJHIHDFXCNFAA-LBPRGKRZSA-O
Synonym r-tert-butyl 3-amino-4-phenylbutanoate, tert-butyl 3r-3-amino-4-phenylbutanoate, 1,1-dimethylethyl 3r-3-amino-4-phenylbutanoate, benzenebutanoic acid, b-amino-, 1,1-dimethylethyl ester, br, t-butyl 3r-3-amino-4-phenylbutanoate, r-tert-butyl3-amino-4-phenylbutanoate, tert-butyl 3r-3-amino-4-phenyl-butanoate, benzenebutanoic acid,b-amino-,1,1-dimethylethyl ester, br
PubChem CID 1501867
IUPAC Name (2S)-4-(tert-butoxy)-4-oxo-1-phenylbutan-2-aminium
SMILES CC(C)(C)OC(=O)C[C@@H]([NH3+])CC1=CC=CC=C1

Specifications

Color White or Yellow
Infrared Spectrum Authentic
Linear Formula (CH3)3COC(O)CH2CH(NH2)CH2C6H5
Quantity 100 mg
Formula Weight 235.3
Percent Purity ≥96.0%
Physical Form Low Melting Solid
Chemical Name or Material tert-Butyl (3R)-3-amino-4-phenylbutanoate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.