accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCl

CAS RN 16606-02-3

IUPAC Name: 2,4',5-trichloro-1,1'-biphenyl
Synonyms: 2,4',5-trichlorobiphenyl
Molecular Weight (g/mol): 257.54
Molecular Formula: C12H7Cl3
InChi Key: VAHKBZSAUKPEOV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results