Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5,6-Tetrachloro-m-xylene Solution, 2000μg/mL in acetone, Ultra Scientific

Catalog No. USIST4401
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USIST4401 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USIST4401 Supplier Agilent Technologies Supplier No. IST4401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16606-02-3 , 67-64-1
Molecular Formula C12H7Cl3
Molecular Weight (g/mol) 257.538
InChI Key VAHKBZSAUKPEOV-UHFFFAOYSA-N
PubChem CID 27960
IUPAC Name 1,4-dichloro-2-(4-chlorophenyl)benzene
SMILES C1=CC(=CC=C1C2=C(C=CC(=C2)Cl)Cl)Cl

Specifications

CAS Max % 99.75, 0.25
Analyte or Component Names Acetone,2,4
Element Acetone,2,4
Solvent or Matrix Acetone
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Chemical Name or Material 2,4,5,6-Tetrachloro-m-xylene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.