accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOCH3O

CAS RN 16630-55-0

IUPAC Name: 3-(methylsulfanyl)propyl acetate
Synonyms: 3-(methylsulfanyl)propyl acetate
Molecular Weight (g/mol): 148.22
Molecular Formula: C6H12O2S
InChi Key: LPZQTNIAYMRVMF-UHFFFAOYSA-N
SMILES: CSCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results