Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)propyl Acetate 98.0+%, TCI America™
SDP

Catalog No. M238625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M238625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M238625G Supplier TCI America Supplier No. M238625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16630-55-0
Molecular Formula C6H12O2S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00053113
InChI Key LPZQTNIAYMRVMF-UHFFFAOYSA-N
Synonym Acetic Acid 3-(Methylthio)propyl Ester
PubChem CID 85519
ChEBI CHEBI:87281
IUPAC Name 3-(methylsulfanyl)propyl acetate
SMILES CSCCCOC(C)=O

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 g
Formula Weight 148.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)propyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.