accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO2H2H2HO2H2H2H

CAS RN 16649-49-3

IUPAC Name: (2,2,2-²H₃)acetyl (²H₃)acetate
Synonyms: (2,2,2-²H₃)acetyl (²H₃)acetate
Molecular Weight (g/mol): 108.13
Molecular Formula: C4H6O3
InChi Key: WFDIJRYMOXRFFG-WFGJKAKNSA-N
SMILES: [2H]C([2H])([2H])C(=O)OC(=O)C([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results