Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic anhydride-d6, for NMR, 98.5 atom % D

Catalog No. AC174670050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174670050 5 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC174670050 Supplier Thermo Scientific Chemicals Supplier No. 174670050
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16649-49-3 | C4H6O3 | 108.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16649-49-3
Molecular Formula C4H6O3
Molecular Weight (g/mol) 108.13
MDL Number MFCD00044225
InChI Key WFDIJRYMOXRFFG-WFGJKAKNSA-N
Synonym acetic anhydride-d6, 2h3 acetic anhydride, acetic anhydride d6, 2h3 acetyl 2h3 acetate, acetic anhydride-d6, 99 atom % d, 2 h? acetyl 2 h? acetate, 2,2,2-trideuterioacetyl 2,2,2-trideuterioacetate
PubChem CID 2723966
IUPAC Name (2,2,2-trideuterioacetyl) 2,2,2-trideuterioacetate
SMILES [2H]C([2H])([2H])C(=O)OC(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Quantity 5 mL
Density 1.1400g/mL
Deuterium (D) 98.0% min.
Linear Formula (CD3CO)2O
Formula Weight 108.14
Grade NMR
Chemical Name or Material Acetic anhydride-d6, 98.5 atom % D
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.