accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16650-14-9

IUPAC Name: 6-bromo-1',3',3'-trimethyl-1',3'-dihydrospiro[chromene-2,2'-indole]
Synonyms: 6-bromo-1',3',3'-trimethylspiro[chromene-2,2'-indole]
Molecular Weight (g/mol): 356.26
Molecular Formula: C19H18BrNO
InChi Key: BDULIJWZMMHIEQ-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C11OC2=C(C=C1)C=C(Br)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results