Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran 98.0+%, TCI America™
SDP

Catalog No. T03701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T03701G Supplier TCI America Supplier No. T03701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16650-14-9
Molecular Formula C19H18BrNO
Molecular Weight (g/mol) 356.263
MDL Number MFCD00059896
InChI Key BDULIJWZMMHIEQ-UHFFFAOYSA-N
Synonym 6-Bromo-1′C,3′C,3′C-trimethylspiro[1(2H)-benzopyran-2,2′C-indoline]
PubChem CID 594671
IUPAC Name 6-bromo-1',3',3'-trimethylspiro[chromene-2,2'-indole]
SMILES CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC(=C4)Br)C)C

Specifications

Quantity 1 g
Formula Weight 356.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,3-Trimethylindolino-6′-bromobenzopyrylospiran [Photochromic Compound]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.