CAS RN 16652-76-9
CAS RN 16652-76-9
IUPAC Name:
4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-methylbutanoate
Synonyms:
benzyl 2-amino-3-methylbutanoate; para-toluene sulfonate
Molecular Weight (g/mol):
379.47
Molecular Formula:
C19H25NO5S
InChi Key:
QWUQVUDPBXFOKF-UHFFFAOYNA-N
SMILES:
CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C(N)C(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Advanced Chem Tech H-Val-OBzl.Tos, Advanced ChemTech
CAS: 16652-76-9 Molecular Formula: C19H25NO5S Molecular Weight (g/mol): 379.47 InChI Key: QWUQVUDPBXFOKF-UHFFFAOYNA-N IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-methylbutanoate SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C(N)C(=O)OCC1=CC=CC=C1