accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3COOOSiCH3CH3H3C

CAS RN 16654-74-3

IUPAC Name: 2,2,10,10-tetramethyl-3,6,9-trioxa-2,10-disilaundecane
Synonyms: 2,2,10,10-tetramethyl-3,6,9-trioxa-2,10-disilaundecane
Molecular Weight (g/mol): 250.49
Molecular Formula: C10H26O3Si2
InChi Key: VZCNNAQIFXLAEZ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OCCOCCO[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results