CAS RN 16655-82-6
IUPAC Name:
3-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl N-methylcarbamate
Synonyms:
3-hydroxycarbofuran
Molecular Weight (g/mol):
237.26
Molecular Formula:
C12H15NO4
InChi Key:
RHSUJRQZTQNSLL-UHFFFAOYNA-N
SMILES:
CNC(=O)OC1=CC=CC2=C1OC(C)(C)C2O
1
–
1
of
1
results
1
3-Hydroxycarbofuran, SPEX CertiPrep™
CAS: 16655-82-6 Molecular Formula: C12H15NO4 Molecular Weight (g/mol): 237.255 InChI Key: RHSUJRQZTQNSLL-UHFFFAOYSA-N PubChem CID: 27975 IUPAC Name: (3-hydroxy-2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate SMILES: CC1(C(C2=C(O1)C(=CC=C2)OC(=O)NC)O)C