accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OHO

CAS RN 1666-04-2

IUPAC Name: 2-hydroxy-3,6-dimethylbenzaldehyde
Synonyms: 2-hydroxy-3,6-dimethylbenzaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: WTHUWXPBPNTSHJ-UHFFFAOYSA-N
SMILES: CC1=C(O)C(C=O)=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results