Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dimethylsalicylaldehyde 98.0+%, TCI America™
SDP

Catalog No. D45075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45075G 5 g
D45071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45075G Supplier TCI America Supplier No. D45075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1666-04-2 | C9H10O2 | 150.177 g/mol

Chemical Identifiers

CAS 1666-04-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD04037328
InChI Key WTHUWXPBPNTSHJ-UHFFFAOYSA-N
Synonym 2-Hydroxy-3,6-dimethylbenzaldehyde
PubChem CID 819297
IUPAC Name 2-hydroxy-3,6-dimethylbenzaldehyde
SMILES CC1=C(C(=C(C=C1)C)O)C=O

Specifications

Melting Point 61°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Chemical Name or Material 3,6-Dimethylsalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.