accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONHN(E)

CAS RN 1666-17-7

IUPAC Name: 4-methyl-N'-[(E)-phenylmethylidene]benzene-1-sulfonohydrazide
Synonyms: 4-methyl-N'-[(E)-phenylmethylidene]benzenesulfonohydrazide
Molecular Weight (g/mol): 274.34
Molecular Formula: C14H14N2O2S
InChi Key: FZFLTDNAHASQQC-RVDMUPIBSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N\N=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results