Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzaldehyde p-Toluenesulfonylhydrazone 98.0+%, TCI America™
SDP

Catalog No. B33255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33255G 5 g
B332525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33255G Supplier TCI America Supplier No. B33255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1666-17-7
Molecular Formula C14H14N2O2S
Molecular Weight (g/mol) 274.338
MDL Number MFCD00009644
InChI Key FZFLTDNAHASQQC-PTNGSMBKSA-N
Synonym Benzaldehyde Tosylhydrazone
PubChem CID 5397192
IUPAC Name N-[(Z)-benzylideneamino]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=CC2=CC=CC=C2

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 274.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzaldehyde p-Toluenesulfonylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.