accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH

CAS RN 1667-12-5

IUPAC Name: [4'-(hydroxymethyl)-[1,1'-biphenyl]-4-yl]methanol
Synonyms: [4'-(hydroxymethyl)-[1,1'-biphenyl]-4-yl]methanol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: SFHGONLFTNHXDX-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CC=C(CO)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results