Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenyldimethanol 98.0+%, TCI America™
SDP

Catalog No. B26235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26235G Supplier TCI America Supplier No. B26235G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1667-12-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00016713
InChI Key SFHGONLFTNHXDX-UHFFFAOYSA-N
Synonym 4,4′C-Bis(hydroxymethyl)biphenyl
PubChem CID 611786
IUPAC Name [4'-(hydroxymethyl)-[1,1'-biphenyl]-4-yl]methanol
SMILES OCC1=CC=C(C=C1)C1=CC=C(CO)C=C1

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldimethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.