accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16670-48-7

IUPAC Name: 2-chloroethyl benzenesulfonate
Synonyms: 2-chloroethyl benzenesulfonate
Molecular Weight (g/mol): 220.67
Molecular Formula: C8H9ClO3S
InChi Key: CIIGWOXXOUVEAD-UHFFFAOYSA-N
SMILES: ClCCOS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results