accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16677-29-5

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-[4-(benzyloxy)phenyl]propanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-[4-(benzyloxy)phenyl]propanoic acid
Molecular Weight (g/mol): 405.45
Molecular Formula: C24H23NO5
InChi Key: IPAODWFPTVIUSZ-ANBDAQEENA-N
SMILES: OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)