CAS RN 166953-64-6
IUPAC Name:
benzyl 4-bromopiperidine-1-carboxylate
Synonyms:
benzyl 4-bromopiperidine-1-carboxylate
Molecular Weight (g/mol):
298.18
Molecular Formula:
C13H16BrNO2
InChi Key:
BFWKSPOSMPKTSV-UHFFFAOYSA-N
SMILES:
BrC1CCN(CC1)C(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-Bromo-N-Z-piperidine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.