accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 166964-33-6

IUPAC Name: 5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride
Synonyms: 5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 281.17
Molecular Formula: C9H6Cl2O2S2
InChi Key: JLTUANWNGKWRQO-UHFFFAOYSA-N
SMILES: CC1=C(SC2=CC=C(Cl)C=C12)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results