Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. MWP00006DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MWP00006DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWP00006DA Supplier Thermo Scientific Chemicals Supplier No. MWP00006DA
Add to Cart

Chemical Identifiers

CAS 166964-33-6
Molecular Formula C9H6Cl2O2S2
Molecular Weight (g/mol) 281.17
MDL Number MFCD00173797
InChI Key JLTUANWNGKWRQO-UHFFFAOYSA-N
Synonym 5-chloro-3-methylbenzo b thiophene-2-sulfonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulphonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulfonyl, 5-chloro-3-methylbenzo b thiophene-2-sulphonylchloride, 5-chloro-3-methyl-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, 5-chloro-3-methyl, zlchem 600, pubchem5100, buttpark 21\07-05, 5-chloro-3-methyl-benzothiophene-2-sulfonyl chloride
PubChem CID 2735756
IUPAC Name 5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride
SMILES CC1=C(SC2=CC=C(Cl)C=C12)S(Cl)(=O)=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 281.18
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.