accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16712-20-2

IUPAC Name: lithium(1+) hydrate chloride
Synonyms: lithium(1+) hydrate chloride
Molecular Weight (g/mol): 60.41
Molecular Formula: ClH2LiO
InChi Key: VXJIMUZIBHBWBV-UHFFFAOYSA-M
SMILES: [Li+].O.[Cl-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results