CAS RN 16750-63-3
IUPAC Name:
magnesium(2+) anisole bromide
Synonyms:
magnesium(2+) anisole bromide
Molecular Weight (g/mol):
211.34
Molecular Formula:
C7H7BrMgO
InChi Key:
FCPRDUXJWIUVPZ-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].COC1=CC=CC=[C-]1
1
–
1
of
1
results
1
Sigma Aldrich 2,2'-Isopropylidenebis[(4S)-4-Tert-Butyl-2-Oxazoline]
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.