accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOH2NOO

CAS RN 167757-45-1

IUPAC Name: benzyl 4-carbamoylpiperidine-1-carboxylate
Synonyms: benzyl 4-carbamoylpiperidine-1-carboxylate
Molecular Weight (g/mol): 262.31
Molecular Formula: C14H18N2O3
InChi Key: UNJWOPBEKPMSGH-UHFFFAOYSA-N
SMILES: NC(=O)C1CCN(CC1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results