Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate, 95%, Thermo Scientific™

Catalog No. CC00094DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00094DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00094DA Supplier Thermo Scientific Chemicals Supplier No. CC00094DA
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 167757-45-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C14H18N2O3
Quantity 1 g
MDL Number MFCD01568760
Synonym benzyl 4-aminocarbonyl tetrahydro-1 2h-pyridinecarboxylate, 1-cbz-4-piperidinecarboxamide, 1-n-cbz-piperidine-4-carboxamide, 1-piperidinecarboxylic acid, 4-aminocarbonyl-, phenylmethyl ester, 4-carbamoyl-piperidine-1-carboxylic acid benzyl ester, n-z-isonipecotinamide, n-z-isonepocotinamide, buttpark 7550-02, ksc532o5b, 1-n-cbz-piperidine-4-carboxyamide
InChI Key UNJWOPBEKPMSGH-UHFFFAOYSA-N
SMILES C1CN(CCC1C(=O)N)C(=O)OCC2=CC=CC=C2
IUPAC Name benzyl 4-carbamoylpiperidine-1-carboxylate
Molecular Weight (g/mol) 262.309
PubChem CID 2776131
Formula Weight 262.31
Percent Purity 95%
Chemical Name or Material Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water fo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.