CAS RN 1678-97-3
IUPAC Name:
1,2,3-trimethylcyclohexane
Synonyms:
cyclohexane, 1,2,3-trimethyl-
Molecular Weight (g/mol):
126.24
Molecular Formula:
C9H18
InChi Key:
DQTVJLHNWPRPPH-UHFFFAOYNA-N
SMILES:
CC1CCCC(C)C1C
1
–
1
of
1
results
1
1,2,3-Trimethylcyclohexane 95.0+%, TCI America™
CAS: 1678-97-3 Molecular Formula: C9H18 Molecular Weight (g/mol): 126.243 MDL Number: MFCD00045496 InChI Key: DQTVJLHNWPRPPH-UHFFFAOYSA-N PubChem CID: 15507 IUPAC Name: 1,2,3-trimethylcyclohexane SMILES: CC1CCCC(C1C)C