Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trimethylcyclohexane 95.0+%, TCI America™
SDP

Catalog No. T08245ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08245ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T08245ML Supplier TCI America Supplier No. T08245ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1678-97-3
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
MDL Number MFCD00045496
InChI Key DQTVJLHNWPRPPH-UHFFFAOYSA-N
PubChem CID 15507
IUPAC Name 1,2,3-trimethylcyclohexane
SMILES CC1CCCC(C1C)C

Specifications

Color Colorless
Boiling Point 151°C
Quantity 5 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Trimethylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.