accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1679-53-4

IUPAC Name: 10-hydroxydecanoic acid
Synonyms: 10-hydroxydecanoic acid 10-OH-capric acid 10-OH-decanoic acid
Molecular Weight (g/mol): 188.27
Molecular Formula: C10H20O3
InChi Key: YJCJVMMDTBEITC-UHFFFAOYSA-N
SMILES: OCCCCCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results