accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16807-13-9

IUPAC Name: 3-iodo-9H-carbazole
Synonyms: 3-iodo-9H-carbazole
Molecular Weight (g/mol): 293.11
Molecular Formula: C12H8IN
InChi Key: OYIGWMXXIFYAGD-UHFFFAOYSA-N
SMILES: IC1=CC2=C(NC3=C2C=CC=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results