Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodocarbazole 98.0+%, TCI America™
SDP

Catalog No. I09195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09195G 5 g
I091925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09195G Supplier TCI America Supplier No. I09195G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16807-13-9
Molecular Formula C12H8IN
Molecular Weight (g/mol) 293.11
MDL Number MFCD00267746
InChI Key OYIGWMXXIFYAGD-UHFFFAOYSA-N
PubChem CID 3678069
IUPAC Name 3-iodo-9H-carbazole
SMILES IC1=CC2=C(NC3=C2C=CC=C3)C=C1

Specifications

Melting Point 199°C
Color White
Quantity 5 g
Formula Weight 293.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.