accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaH2NNHNOOHSOOOSOOO(E)

CAS RN 1681-60-3

IUPAC Name: disodium (3E)-3-[2-(4-aminophenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (3E)-3-[2-(4-aminophenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 483.38
Molecular Formula: C16H11N3Na2O8S2
InChi Key: BRJHCEZUOXQKTH-AXHWYJOWSA-L
SMILES: [Na+].[Na+].NC1=CC=C(N\N=C2/C(=O)C3=C(O)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C=C1
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results