Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Violet 3, TCI America™
SDP

Catalog No. P059225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P059225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P059225G Supplier TCI America Supplier No. P059225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1681-60-3
Molecular Formula C16H11N3Na2O8S2
Molecular Weight (g/mol) 483.38
MDL Number MFCD00049380
InChI Key BRJHCEZUOXQKTH-AXHWYJOWSA-L
Synonym 3-(4-Aminophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, Victoria Violet, Disodium 3-(4-Aminophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonate, Pontacyl Violet 4BSN
PubChem CID 44135350
IUPAC Name disodium (3E)-3-[2-(4-aminophenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].NC1=CC=C(N\N=C2/C(=O)C3=C(O)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C=C1

Specifications

Color Black/Purple
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 483.38
Chemical Name or Material Acid Violet 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.