accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1682-20-8

IUPAC Name: tetrafluoropyridin-4-amine
Synonyms: tetrafluoropyridin-4-amine
Molecular Weight (g/mol): 166.08
Molecular Formula: C5H2F4N2
InChi Key: PLVFNMQHMRRBAA-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=NC(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)