Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,3,5,6-tetrafluoropyridine 97.0+%, TCI America™
SDP

Catalog No. A15461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15461G Supplier TCI America Supplier No. A15461G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorinated building block with a minimum purity of 97.0% suitable for laboratory research.

Chemical Identifiers

CAS 1682-20-8
Molecular Formula C5H2F4N2
Molecular Weight (g/mol) 166.079
MDL Number MFCD00010855
InChI Key PLVFNMQHMRRBAA-UHFFFAOYSA-N
Synonym 4-amino-2,3,5,6-tetrafluoropyridine, 4-aminotetrafluoropyridine, tetrafluoropyridin-4-amine, 2,3,5,6-tetrafluoro-4-pyridinamine, 2,3,5,6-tetrafluoro-4-pyridylamine, pubchem2992, acmc-1bzbr, 4-amino-tetrafluoropyridine, 4-amino-2,3,5,6-tetrafluorropyridine, 4-pyridinamine, 2,3,5,6-tetrafluoro
PubChem CID 1270911
IUPAC Name 2,3,5,6-tetrafluoropyridin-4-amine
SMILES C1(=C(C(=NC(=C1F)F)F)F)N

Specifications

Melting Point 86°C
Quantity 1 g
Formula Weight 166.08
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.