accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NH3HOOO

CAS RN 16820-18-1

IUPAC Name: (2S)-2-azaniumyl-3-hydroxy-2-methylpropanoate
Synonyms: (2S)-2-ammonio-3-hydroxy-2-methylpropanoate
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: CDUUKBXTEOFITR-BYPYZUCNSA-N
SMILES: C[C@]([NH3+])(CO)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results