Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (+)-2-Methyl-L-serine, 99%

Catalog No. AC290012500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290012500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290012500 Supplier Thermo Scientific Chemicals Supplier No. 290012500
Add to Cart

CAS: 16820-18-1 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16820-18-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00145243
InChI Key CDUUKBXTEOFITR-BYPYZUCNSA-N
Synonym 2-methyl-l-serine, +-2-methyl-l-serine, s-2-amino-3-hydroxy-2-methylpropanoic acid, l-, a-methylserine, l-2-methylserine, s-2-methylserine, 2s-2-amino-3-hydroxy-2-methylpropanoic acid, serine, 2-methyl, alpha-methyl-l-serine, alpha-methyl-l-ser
PubChem CID 7000050
ChEBI CHEBI:74819
IUPAC Name (2S)-2-amino-3-hydroxy-2-methylpropanoic acid
SMILES CC(CO)(C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 286.0°C to 288.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOCH2C(CH3)NH2COOH
Quantity 250 mg
Specific Rotation + 3.30
Solubility Information Solubility in water: soluble
Formula Weight 119.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (+)-2-Methyl-L-serine, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.