accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16820-54-5

IUPAC Name: (4-methoxynaphthalen-1-yl)methanol
Synonyms: (4-methoxynaphthalen-1-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: VBXSDYNXURBEQH-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=C(CO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results